missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chromate, 2% w/v, Reagents
Sodium Chromate, 2% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CS1449001A
Specifications
| 100°C | |
| Yellow | |
| Liquid | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 2% | |
| Poly Bottle |
| Sodium Chromate, 2% w/v | |
| 7732-18-5,10034-82-9 | |
| 1 L | |
| Chromium disodium oxide | |
| O | |
| 18.02 | |
| Reagent | |
| 1.02 |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Chromium disodium oxide | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |