missing translation for 'onlineSavingsMsg'
Learn More

Sodium cholate hydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1707409

 View more versions of this product

Catalog No. AAA1707409

Only null left
Explore available promotions

Description

Description

Biochemical solubilizing agent Sodium cholate hydrate is a non-denaturing detergent used for the extraction of membrane proteins. It is involved in the study to assess infiltration of pressurized promoter solutions in a mesoporous silica. Further, it serves as a biochemical solubilizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biochemical solubilizing agent Sodium cholate hydrate is a non-denaturing detergent used for the extraction of membrane proteins. It is involved in the study to assess infiltration of pressurized promoter solutions in a mesoporous silica. Further, it serves as a biochemical solubilizing agent.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong acids.
Specifications

Specifications

Sodium cholate hydrate
C24H39NaO5
10 g
sodium cholate hydrate, a-cholan-24-oic acid sodium salt, c24h39o5.na.h2o, sodium cholate hydrate, bioxtra, sodium cholate hydrate, pharmagrade, manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., cholalic acid sodium salt, 3, sodium cholate hydrate, bioxtra, sodium cholate hydrate dried material, nt, a-trihydroxy-5, 3, a,7, a,12, a-trihydroxy-5
NRHMKIHPTBHXPF-TUJRSCDTSA-M
[Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C
430.56
430.56 (Anhydrous)
206986-87-0
MFCD00064138 MFCD00003672
3582354
Soluble in water.
+32° (c=0.5 in Water)
sodium;(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate;hydrate
23679061
99%

Chemical Identifiers

206986-87-0
430.56
NRHMKIHPTBHXPF-TUJRSCDTSA-M
sodium;(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate;hydrate
C24H39NaO5
MFCD00064138 MFCD00003672
23679061
[Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : FZ9600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only