accessibility menu, dialog, popup

UserName

CAS RN 206986-87-0

IUPAC Name: sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
Synonyms: sodium cholate|cholic acid, monosodium salt
Molecular Weight (g/mol): 430.56
Molecular Formula: C24H39NaO5
InChi Key: NRHMKIHPTBHXPF-TUJRSCDTSA-M
SMILES: [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results