missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium cholate hydrate, 99%, Thermo Scientific Chemicals
Description
Biochemical solubilizing agent Sodium cholate hydrate is a non-denaturing detergent used for the extraction of membrane proteins. It is involved in the study to assess infiltration of pressurized promoter solutions in a mesoporous silica. Further, it serves as a biochemical solubilizing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Biochemical solubilizing agent Sodium cholate hydrate is a non-denaturing detergent used for the extraction of membrane proteins. It is involved in the study to assess infiltration of pressurized promoter solutions in a mesoporous silica. Further, it serves as a biochemical solubilizing agent.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong acids.
Biochemical solubilizing agent Sodium cholate hydrate is a non-denaturing detergent used for the extraction of membrane proteins. It is involved in the study to assess infiltration of pressurized promoter solutions in a mesoporous silica. Further, it serves as a biochemical solubilizing agent.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong acids.
Specifications
Specifications
| Quantity | 10 g |
| Beilstein | 3582354 |
| Synonym | sodium cholate hydrate, cholalic acid sodium salt, c24h39o5.na.h2o, sodium cholate hydrate, bioxtra, sodium cholate hydrate dried material, nt, sodium cholate hydrate, from ox and/or sheep bile, 3, a,7, a,12, a-trihydroxy-5, a-cholan-24-oic acid sodium salt, sodium cholate hydrate, suitable for cell culture, bioreagent, 4r-4-1s,2s,7s,11s,16s,5r,9r,10r,14r,15r-5,9,16-trihydroxy-2,15-dimethylte tracyclo 8.7.0.0<2,7>.0<11,15*gt; heptadec-14-yl pentanoic acid, sodium salt, hyd rate, sodium cholate hydrate, pharmagrade, manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production. |
| Solubility Information | Soluble in water. |
| Optical Rotation | +32° (c=0.5 in Water) |
| Formula Weight | 430.56 (Anhydrous) |
| Percent Purity | 99% |
| Chemical Name or Material | Sodium cholate hydrate |
Safety and Handling
| RTECSNumber | FZ9600000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only