missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, Saturated 32.5% (w/v), Ricca Chemical

Catalog No. 7220532
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7220532 1 L Natural Poly Bottle
722051 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7220532 Supplier Ricca Chemical Company Supplier No. 7220.532
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Color Colorless
CAS Min % 74.0, 24.05
CAS Max % 77.0, 25.03
Physical Form Liquid
Quantity 1 L
Percent Purity 32.5% W/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 32.5% (w/v)
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Chloride, Saturated
ShelfLife 24 months