missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, Powder, USP, 99-100.5%, Spectrum™ Chemical

SO160, 7647-14-5, NaCl

Supplier:  Spectrum Chemical Mfg Cor SO16025KGCS4

 View more versions of this product

Catalog No. 18606425


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Sodium Chloride, Powder, USP is used in the formulation of oral hygiene products, shampoos, fragrance, skin, hair, nail, cleansing, suntan, makeup and bath products. This grade is specifically used in oral care applications. This product is not sterile and not intended for use in the manufacture of injectable dosage forms, peritoneal dialysis solutions, hemodialysis solutions or hemofiltration solutions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

7647-14-5
4 x 2.5 kg
MFCD00003477
[Na+].[Cl-]
58.44
0.005
1
ClNa
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
99 to 100.5%
Poly Bottle

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
[Na+].[Cl-]
ClNa
MFCD00003477
sodium chloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions