missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, Powder, USP, 99-100.5%, Spectrum™ Chemical

Catalog No. p-7158200
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
12 kg
2.5 kg
4 x 2.5 kg
50 kg
500 g
6 x 500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18606422 500 g Poly Bottle
18606424 2.5 kg Poly Bottle
18606423 6 x 500 g Poly Bottle
18606425 4 x 2.5 kg Poly Bottle
18606426 12 kg Poly Pail
18606427 50 kg Fiber Drum
6 options

Catalog No. 18606422

Supplier: Spectrum Chemical Mfg Cor SO160500GM

SO160, 7647-14-5, NaCl

Spectrum™ Chemical Sodium Chloride, Powder, USP is used in the formulation of oral hygiene products, shampoos, fragrance, skin, hair, nail, cleansing, suntan, makeup and bath products. This grade is specifically used in oral care applications. This product is not sterile and not intended for use in the manufacture of injectable dosage forms, peritoneal dialysis solutions, hemodialysis solutions or hemofiltration solutions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99 to 100.5%
Grade USP
Loss on Drying 0.005
Packaging Poly Bottle