missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, Non-Sterile, Certified, 0.850% (w/v) ±0.005%, LabChem™

Catalog No. p-5388242
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC234501 500 mL
LC234502 1 L
LC234504 4 L
LC234505 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. LC234501 Supplier LabChem Supplier No. LC234501
Only null left

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Color Colorless
CAS Max % 99.15, 0.85
Physical Form Liquid
Quantity 500 mL
Linear Formula NaCl
Solubility Information Soluble in water
Formula Weight 58.44
Percent Purity 0.85% ±0.005% (w/v)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen chloride
Chemical Name or Material Sodium Chloride, Non-Sterile
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature