Learn More
Gibco™ Sodium Chloride (NaCl) Solutions, 0.1–2.0 M
Gibco Sodium Chloride (NaCl) solutions are sterile-filtered, packaged in bioprocess containers, and ready-to-use. Sourcing NaCl solutions can help streamline your buffer preparation allowing you to focus on additional critical steps in your biopharmaceutical processes.
Supplier: Gibco™ A5792202
Description
We mix compendial-grade sodium chloride (NaCl) with the same bulk water used in our Gibco Water for Injection (WFI) for Cell Culture to produce solutions that meet the most stringent expectations of the pharmaceutical, biopharma, and diagnostic industries. NaCl solutions are manufactured to cGMP (21CFR820) and ISO 13485 standards, promoting consistency and convenience for every stage of the bioprocessing pipeline.
Product features:
• Multiple concentrations available (0.1 M, 0.2 M, 0.5 M, 1.0 M, 2.0 M)
• Available in a range of pack sizes (5 L, 20 L, 200 L)
• Established products with set specifications are ready to order
• Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
• Readily accessible documentation for accelerated qualification
User benefits:
• Eases the time requirements dedicated for in-house buffer preparation
• Mitigates space required for equipment storage
• Streamlines in-house chromatography processes related to formulation, hydration, testing, and validation
• Meets workflow needs for all stages of biologic development and manufacturing
Specifications
| Sodium Chloride | |
| 0.1 M | |
| Liquid | |
| 0.1 M NaCl | |
| Sterile-filtered | |
| 0.2 μm Sterile-filtered | |
| 167–207 mOsm/kg |
| Clear | |
| 4.5–8.0 | |
| 20 L | |
| Tote | |
| GMP - 21 CFR 820, ISO 13485 | |
| ≤ 1.0 EU/mL |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.