missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Sodium Chloride (NaCl) Solutions, 0.1–2.0 M

Catalog No. A5792201
Change view
Click to view available options
Quantity:
5 L
20 L
200 L
Packaging:
Drum
Sample
Tote
Concentration:
0.1 M
0.2 M
0.5 M
1.0 M
2.0 M
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Concentration
A5792201 5 L Sample 0.1 M
A5792202 20 L Tote 0.1 M
A5792203 200 L Drum 0.1 M
A5802801 5 L Sample 0.2 M
A5802802 20 L Tote 0.2 M
A5802803 200 L Drum 0.2 M
A5802901 5 L Sample 0.5 M
A5802902 20 L Tote 0.5 M
A5802903 200 L Drum 0.5 M
A5803001 5 L Sample 1.0 M
A5803002 20 L Tote 1.0 M
A5803003 200 L Drum 1.0 M
A5803101 5 L Sample 2.0 M
A5803102 20 L Tote 2.0 M
A5803103 200 L Drum 2.0 M
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. A5792201 Supplier Gibco™ Supplier No. A5792201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gibco Sodium Chloride (NaCl) solutions are sterile-filtered, packaged in bioprocess containers, and ready-to-use. Sourcing NaCl solutions can help streamline your buffer preparation allowing you to focus on additional critical steps in your biopharmaceutical processes.

We mix compendial-grade sodium chloride (NaCl) with the same bulk water used in our Gibco Water for Injection (WFI) for Cell Culture to produce solutions that meet the most stringent expectations of the pharmaceutical, biopharma, and diagnostic industries. NaCl solutions are manufactured to cGMP (21CFR820) and ISO 13485 standards, promoting consistency and convenience for every stage of the bioprocessing pipeline.

Product features:
• Multiple concentrations available (0.1 M, 0.2 M, 0.5 M, 1.0 M, 2.0 M)
• Available in a range of pack sizes (5 L, 20 L, 200 L)
• Established products with set specifications are ready to order
• Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
• Readily accessible documentation for accelerated qualification

User benefits:
• Eases the time requirements dedicated for in-house buffer preparation
• Mitigates space required for equipment storage
• Streamlines in-house chromatography processes related to formulation, hydration, testing, and validation
• Meets workflow needs for all stages of biologic development and manufacturing

Specifications

Chemical Name or Material Sodium Chloride
Color Clear
Concentration 0.1 M
pH 4.5–8.0
Physical Form Liquid
Quantity 5 L
Formulation 0.1 M NaCl
Packaging Sample
Sterility Sterile-filtered
Certifications/Compliance GMP - 21 CFR 820, ISO 13485
Sterilization Method 0.2 μm Sterile-filtered
Endotoxin Level ≤ 1.0 EU/mL
Osmolality 167–207 mOsm/kg
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.