Learn More
Sodium Chloride (Granular/USP/FCC), Fisher Chemical™
Supplier: Fisher Chemical S64050
Specifications
| 1461°C | |
| 801°C | |
| White | |
| Solid | |
| ClNa | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| 99.0 to 100.5% (USP) | |
| Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC) | |
| Fiber Drum | |
| 2.165g/cm³ |
| Sodium Chloride | |
| 7647-14-5 | |
| 5.0 to 8.0 | |
| 50 kg | |
| MFCD00003477 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| 58.44 | |
| FCC/USP | |
| 0.5% max. (FCC), 0.5% max. (USP), 0.5% max. (USP/FCC) | |
| 1mmHg at 440°C |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| 5234 | |
| sodium chloride |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:1
Flammability:0
Instability:1