missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride (Granular/USP/FCC), Fisher Chemical™

Catalog No. p-33749
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
50 kg
350 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S640500 500 g Poly Bottle
S640350LB 350 lb. Fiber Drum
S6403 3 kg Poly Bottle
S64010 10 kg Poly Pail
S64050 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S640500 Supplier Thermo Fisher Scientific Supplier No. S640 500
Only null left

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461°C
Melting Point 801°C
Color White
pH 5.0 to 8.0
Physical Form Solid
Quantity 500 g
Formula Weight 58.44
Percent Purity 99.0 to 100.5% (FCC)
Grade FCC/USP
Identification Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC)
Loss on Drying 0.5% max. (FCC), 0.5% max. (USP), 0.5% max. (USP/FCC)
Packaging Poly Bottle
Vapor Pressure 1mmHg at 440°C
Density 2.165g/cm³
Chemical Name or Material Sodium Chloride
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1