missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride (Granular/USP/FCC), Fisher Chemical™

Supplier:  Fisher Chemical S6403

 View more versions of this product

Catalog No. S640-3

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1461°C
801°C
White
Solid
ClNa
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
[Na+].[Cl-]
58.44
CHEBI:26710
99.0 to 100.5% (FCC)
Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC)
Poly Bottle
2.165g/cm³
Sodium Chloride
7647-14-5
5.0 to 8.0
3 kg
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
5234
58.44
FCC/USP
0.5% max. (FCC), 0.5% max. (USP), 0.5% max. (USP/FCC)
1mmHg at 440°C

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
sodium chloride
ClNa
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
CHEBI:26710
[Na+].[Cl-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1

SDS Documents

Documents

Promotions

Promotions