missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, For Chloride, Certified, 0.0250N ±0.0005N (0.025M), LabChem™
Supplier: LabChem LC235501
Specifications
| Sodium Chloride | |
| For Chloride | |
| 99.85,0.15 | |
| NaCl | |
| 0.0250N ±0.0005N (0.025M) | |
| Soluble in water | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| Certified | |
| Poly Bottle | |
| Hydrogen chloride | |
| Liquid |
| Colorless | |
| 7647-14-5,7732-18-5 | |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| 58.44 | |
| Passes Test | |
| Traceable to NIST | |
| 500 mL |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| 5234 | |
| sodium chloride |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature