missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, Certified, 25% (w/v) ±1%, LabChem™

Catalog No. LC235201
Change view
Click to view available options
Quantity:
500 mL
4 L
Catalog No. Quantity
LC235201 500 mL
LC235204 4 L
2 options

Catalog No. LC235201

Supplier: LabChem LC235201

Only null left

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Color Colorless
CAS Max % 79.21, 20.79
Physical Form Liquid
Quantity 500 mL
Linear Formula NaCl
Solubility Information Soluble in water
Formula Weight 58.44
Percent Purity 25% ±1% (w/v)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.15g/mL
Decomposition Information Hydrogen chloride
Density 1.15g/mL
Chemical Name or Material Sodium Chloride
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature