missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, Certified, 1.000N ±0.005N (1.0M), LabChem™

Catalog No. LC235802
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC235802 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC235802 Supplier LabChem Supplier No. LC235802
Only null left

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Color Colorless
CAS Max % 94.4, 5.6
Physical Form Liquid
Quantity 1 L
Linear Formula NaCl
Solubility Information Soluble in water
Formula Weight 58.44
Grade Certified
Concentration or Composition (by Analyte or Components) 1.000N ±0.005N (1.0M)
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.04g/mL
Decomposition Information Hydrogen chloride
Density 1.04g/mL
Chemical Name or Material Sodium Chloride
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature