missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, ACS Reagent, Reagents

Sodium Chloride, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

$131.70 - $4183.00

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym Table salt
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-337-744
View Documents
Labchem Inc
C131300010KG
10 kg
Each for $572.77
Only null left
 
01-337-743
View Documents
Labchem Inc
C131300012KG
12 kg
Each for $520.83
Only null left
 
01-337-742
View Documents
Labchem Inc
C131300025A5
2.5 kg
Each for $304.20
Only null left
 
01-337-741
View Documents
Labchem Inc
C1313000350F5
350 lb.
Each for $4,183.00
Only null left
 
01-337-740
View Documents
Labchem Inc
C1313000500A5
500 g
Each for $131.70
Only null left
 
01-337-739
View Documents
Labchem Inc
C131300050F4
50 kg
Each for $2,378.77
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
ClNa
MFCD00003477
Table salt
[Na+].[Cl-]

Specifications

1461°C
Colorless/White
6.7 to 7.3
10 kg
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
ACS
2.17
801°C
7647-14-5
Solid
ClNa
Table salt
[Na+].[Cl-]
58.44
Poly Pail
Sodium Chloride, ACS Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'