missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, ACS Reagent, Reagents
Sodium Chloride, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C131300050F4
Specifications
| 1461°C | |
| 801°C | |
| 7647-14-5 | |
| Solid | |
| ClNa | |
| Table salt | |
| [Na+].[Cl-] | |
| 58.44 | |
| Fiber Drum |
| Sodium Chloride, ACS Reagent | |
| Colorless/White | |
| 6.7 to 7.3 | |
| 50 kg | |
| MFCD00003477 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| ACS | |
| 2.17 |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride |
| ClNa | |
| MFCD00003477 | |
| Table salt | |
| [Na+].[Cl-] |