missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Sodium Chloride, ACS Reagent Grade, Calbiochem™,
$62.70 - $105.00
Chemical Identifiers
| CAS | 7647-14-5 |
|---|---|
| Molecular Formula | ClNa |
| Molecular Weight (g/mol) | 58.44 |
| MDL Number | MFCD00003477 |
| InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
| Synonym | NaCl |
| PubChem CID | 5234 |
| ChEBI | CHEBI:26710 |
| IUPAC Name | sodium chloride |
| SMILES | [Na+].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
567440500GM
|
MilliporeSigma™
567440500GM |
500 g | Glass Bottle |
Each for $62.70
|
|
||||
|
5674401KG
|
MilliporeSigma™
5674401KG |
1 kg | Glass Bottle |
Each for $105.00
|
|
||||
Chemical Identifiers
| ClNa | |
| MFCD00003477 | |
| NaCl | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
Specifications
| White | |
| ClNa | |
| NaCl | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| ACS Reagent |
| 7647-14-5 | |
| MFCD00003477 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| ≥99% |