missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Sodium Chloride, ACS Reagent Grade, Calbiochem™,

Catalog No. 567440500GM
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
567440500GM 500 g Glass Bottle
5674401KG 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 567440500GM Supplier MilliporeSigma™ Supplier No. 567440500GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym NaCl
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Color White
Quantity 500 g
Packaging Glass Bottle
Percent Purity ≥99%
Grade ACS Reagent