missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Sodium Chloride, ACS Reagent Grade, Calbiochem™,
Supplier: MilliporeSigma™ 5674401KG
Specifications
| White | |
| 1 kg | |
| ClNa | |
| NaCl | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| ACS Reagent |
| 7647-14-5 | |
| Glass Bottle | |
| MFCD00003477 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| ≥99% |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| 5234 | |
| sodium chloride |
| ClNa | |
| MFCD00003477 | |
| NaCl | |
| CHEBI:26710 | |
| [Na+].[Cl-] |