missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, ACS, ≥99.5%, Honeywell™ Fluka™
Free-flowing, ACS reagent, anhydrous,≥99%
$447.97 - $447.97
Chemical Identifiers
| CAS | 7647-14-5 |
|---|---|
| Molecular Formula | ClNa |
| Molecular Weight (g/mol) | 58.44 |
| MDL Number | MFCD00003477 |
| InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
| Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
| PubChem CID | 5234 |
| ChEBI | CHEBI:26710 |
| SMILES | [Na+].[Cl-] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| 5234 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| CHEBI:26710 |
Specifications
| 1465°C | |
| 7647-14-5 | |
| 7.0 (at 20°C) | |
| 2.5 kg | |
| NaCl | |
| NONH for all modes of transport | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium;chloride | |
| 5234 | |
| 58.44g/mol | |
| Odorless | |
| 2.160g/cm3 at 20°C |
| 800°C | |
| Colorless | |
| Crystalline | |
| ClNa | |
| MFCD00003477 | |
| 3534976 | |
| 358g/L at 20°C | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| ≥99% | |
| Fiberboard box | |
| Sodium chloride |
Safety and Handling
RTECSNumber : 231-598-3