missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, ACS, ≥99.5%, Honeywell™ Fluka™

Catalog No. 60047480
Change view
Click to view available options
Quantity:
2.5 kg
Catalog No. Quantity
60047480 2.5 kg
1 options

Catalog No. 60047480

Supplier: Honeywell Chemicals 7463982.5KG

Only null left

Free-flowing, ACS reagent, anhydrous,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1465°C
Melting Point 800°C
Color Colorless
pH 7.0 (at 20°C)
Physical Form Crystalline
Quantity 2.5 kg
Linear Formula NaCl
UN Number NONH for all modes of transport
Beilstein 3534976
Solubility Information 358g/L at 20°C
IUPAC Name sodium;chloride
Formula Weight 58.44g/mol
Percent Purity ≥99%
Odor Odorless
Packaging Fiberboard box
Density 2.160g/cm3 at 20°C
Chemical Name or Material Sodium chloride
Show More Show Less
RTECSNumber 231-598-3