missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sodium chloride, 99.5%, for biochemistry
CAS: 7647-14-5 | ClNa | 58.44 g/mol
Supplier: Thermo Scientific Chemicals 447300050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1461.0°C | |
| 801.0°C | |
| 5 kg | |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| 58.44 | |
| Biochemical |
| Sodium chloride | |
| 7647-14-5 | |
| 99.4% min. (Argentometry) | |
| NaCl | |
| 15,8734 | |
| (0.1 M in water) Clear colorless | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| 99.5% | |
| Plastic bottle |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| 5234 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| CHEBI:26710 |
Safety and Handling
EINECSNumber : 231-598-3
RUO – Research Use Only