missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium chloride, 99.5%, for biochemistry

Catalog No. p-4813647
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447302500 250 g Plastic bottle
AC447300010 1 kg Plastic bottle
AC447300050 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC447302500 Supplier Thermo Scientific Chemicals Supplier No. 447302500
Only null left

CAS: 7647-14-5 | ClNa | 58.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461.0°C
Melting Point 801.0°C
Quantity 250 g
Assay Percent Range 99.4% min. (Argentometry)
Linear Formula NaCl
Merck Index 15,8734
Solubility Information (0.1 M in water) Clear colorless
IUPAC Name sodium chloride
Formula Weight 58.44
Percent Purity 99.5%
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Sodium chloride
Show More Show Less
EINECSNumber 231-598-3

RUO – Research Use Only