missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, 3% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company R72137004A

Catalog No. R72137004A


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Chloride
3% (w/v), Aqueous Solution
3.00 ± 0.03% (w/v)
MFCD00003477
Solubility in Water: Infinite
[Na+].[Cl-]
58.44
CHEBI:26710
Laboratory
Natural Poly Bottle
6.7 to 7.3
Liquid
Colorless
7647-14-5,7732-18-5
ClNa
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
5234
3%
Odorless
Aqueous Solution
4 L

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
sodium chloride
ClNa
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
CHEBI:26710
[Na+].[Cl-]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions