missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, 1.00 M, Ricca Chemical

Catalog No. p-3340319
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7227132 1 L Natural Poly Bottle
722711 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7227132 Supplier Ricca Chemical Company Supplier No. 7227.132
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Color Colorless
CAS Min % 93.0, 5.41
CAS Max % 96.0, 5.64
Physical Form Liquid
Quantity 1 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1.00 Molar
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Chloride
ShelfLife 24 months