missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, 1.0% (w/v), Ricca Chemical
$172.25 - $302.31
Chemical Identifiers
| CAS | 7647-14-5 |
|---|---|
| Molecular Formula | ClNa |
| Molecular Weight (g/mol) | 58.44 |
| MDL Number | MFCD00003477 |
| InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
| Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
| PubChem CID | 5234 |
| ChEBI | CHEBI:26710 |
| IUPAC Name | sodium chloride |
| SMILES | [Na+].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R7212500500
|
Ricca Chemical Company
R7212500500A |
500 mL | Natural Poly Bottle |
Each for $172.25
|
|
||||
|
R72125001A
|
Ricca Chemical Company
R72125001A |
1 L | Natural Poly Bottle |
Each for $302.31
|
|
||||
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| 5234 | |
| sodium chloride |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
Specifications
| Colorless | |
| 1.00 ± 0.01% (w/v) | |
| MFCD00003477 | |
| Solubility in Water: Infinite | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| Laboratory | |
| Natural Poly Bottle | |
| 6.7 to 7.3 | |
| Liquid |
| 7647-14-5,7732-18-5 | |
| ClNa | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| 1% | |
| Odorless | |
| Aqueous Solution | |
| 500 mL | |
| Sodium Chloride, 1.0% (w/v), Aqueous Solution |
Safety and Handling
ShelfLife : 24 months
Recommended Storage : Ambient