missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride 0.9% (Normal Saline), USP, Sterile Grade, Cytiva Hyclone™

Catalog No. p-7161881
Change view
Click to view available options
Quantity:
6 x 500 mL
20 L
6 x 1 L
Packaging:
Bioprocess Bag
PET Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
Z1376 6 x 500 mL PET Plastic Bottle
Z1377 6 x 1 L PET Plastic Bottle
Z1379 20 L Bioprocess Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. Z1376 Supplier Cytiva Supplier No. Z1376
Only null left

Sodium Chloride Injection, USP is a sterile solution of sodium chloride in water for injection. It contains no antimicrobial agents. It contains NLT 95.0% and NMT 105.0% of the labeled amount of sodium chloride (NaCl).

  • Manufactured under cGMP and ISO 13485 guidelines in a FDA registered facility
  • Sterile and endotoxin tested
  • Ideal for medical device and biopharmaceutical manufacturing
  • Suitable in research, testing, cell culture and molecular protocols

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Quantity 6 x 500 mL
Concentration or Composition (by Analyte or Components) 0.009
Packaging PET Plastic Bottle
Grade USP
ShelfLife 18 Months
Recommended Storage 15-30°C