missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, 0.5078 N, Conductivity = 47,600 μS/cm at 25°C, Ricca Chemical

$108.72 - $108.72

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
7225-16
View Documents
Ricca Chemical Company
722516
500 mL Natural Poly Bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
sodium chloride
ClNa
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
CHEBI:26710
[Na+].[Cl-]

Specifications

Natural Poly Bottle
APHA for Salinity
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
Conductivity Standard
[Na+].[Cl-]
58.44
5234
0.5078N
7647-14-5,7732-18-5
ClNa
Approximately 0°C
Conductivity Standard
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
500 mL
CHEBI:26710
Sodium Chloride
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'