missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, 0.0250 N (N/40), Ricca Chemical
Supplier: Ricca Chemical Company 722216
Specifications
| Sodium Chloride | |
| ClNa | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| 0.0250N (N/40) | |
| 6.7 to 7.3 |
| 7647-14-5,7732-18-5 | |
| MFCD00003477 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| Laboratory | |
| Natural Poly Bottle | |
| 500 mL |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| 5234 | |
| sodium chloride |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| CHEBI:26710 | |
| [Na+].[Cl-] |