missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., anhydrous, ≥99.8% (calc. to the dried substance), Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., anhydrous,≥99.8% (calc. to the dried substance)
$141.55 - $2488.26
Chemical Identifiers
| CAS | 497-19-8 |
|---|---|
| Molecular Formula | CNa2O3 |
| Molecular Weight (g/mol) | 105.99 |
| MDL Number | MFCD00003494 |
| InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
| Synonym | sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda |
| PubChem CID | 10340 |
| ChEBI | CHEBI:29377 |
| SMILES | [Na+].[Na+].[O-]C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
6002352
|
Honeywell-Fluka
31432250G |
250 g |
Each for $141.55
|
|
|||||
|
6002354
|
Honeywell-Fluka
31432500G |
500 g |
Each for $327.45
|
|
|||||
|
6002350
|
Honeywell-Fluka
314321KG |
1 kg |
Each for $471.29
|
|
|||||
|
6002351
|
Honeywell-Fluka
3143225KG |
2.5 kg |
Each for $1,044.13
|
|
|||||
|
6002355
|
Honeywell-Fluka
314325KG |
5 kg |
Each for $1,280.68
|
|
|||||
|
60045946
|
Honeywell-Fluka
314326X1KG |
6 x 1 kg |
Case of 6 Each for $2,488.26
|
|
|||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 497-19-8 | |
| 105.99 | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| 10340 | |
| [Na+].[Na+].[O-]C([O-])=O |
| CNa2O3 | |
| MFCD00003494 | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| CHEBI:29377 |
Specifications
| 497-19-8 | |
| 250 g | |
| Na2CO3 | |
| NONH for all modes of transport | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| [Na+].[Na+].[O-]C([O-])=O | |
| 105.99 | |
| CHEBI:29377 | |
| ≥99.8% (calc. to the dried substance) | |
| Sodium Carbonate, Reag. Ph. Eur. reag. ISO |
| Anhydrous | |
| CNa2O3 | |
| MFCD00003494 | |
| 4154566 | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| disodium;carbonate | |
| 10340 | |
| 105.99g/mol | |
| ACS/puriss. p.a. |
Safety and Handling
ShelfLife : 1260 days from date of manufacture
EINECSNumber : 207-838-8
Recommended Storage : Room Temp