missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., anhydrous, ≥99.8% (calc. to the dried substance), Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., anhydrous,≥99.8% (calc. to the dried substance)
Supplier: Honeywell Chemicals 314325KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Sodium Carbonate | |
| 497-19-8 | |
| 5 kg | |
| Na2CO3 | |
| NONH for all modes of transport | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| [Na+].[Na+].[O-]C([O-])=O | |
| 105.99 | |
| CHEBI:29377 | |
| ≥99.8% (calc. to the dried substance) |
| Reag. Ph. Eur. reag. ISO | |
| Anhydrous | |
| CNa2O3 | |
| MFCD00003494 | |
| 4154566 | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| disodium carbonate | |
| 10340 | |
| 105.99g/mol | |
| ACS/puriss. p.a. |
Chemical Identifiers
| 497-19-8 | |
| 105.99 | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| 10340 | |
| [Na+].[Na+].[O-]C([O-])=O |
| CNa2O3 | |
| MFCD00003494 | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| CHEBI:29377 |
Safety and Handling
P305 + P351 + P338
H319
EINECSNumber : 207-838-8
RTECSNumber : VZ4050000