missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., anhydrous, ≥99.8% (calc. to the dried substance), Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Melting Point | 851°C (literature) |
| CAS Min % | ≥90.0000% |
| CAS Max % | ≤100.0000% |
| Physical Form | Anhydrous |
| Quantity | 6 x 1 kg |
| Linear Formula | Na2CO3 |
| Beilstein | 4154566 |
| IUPAC Name | disodium;carbonate |
| Formula Weight | 105.99 |
| Percent Purity | >99.8% |
| Show More |
Safety and Handling
| ShelfLife | 1260 days from date of manufacture |
| EINECSNumber | 207-838-8 |
| Recommended Storage | Room Temp |