missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate, Hydrous, NF, 99.5-100.5%, Spectrum™ Chemical
SO150, 5968-11-6, Na2CO3·H2O
$548.96 - $6470.18
Chemical Identifiers
| CAS | 5968-11-6 |
|---|---|
| Molecular Formula | CH2Na2O4 |
| Molecular Weight (g/mol) | 124.00 |
| InChI Key | MQRJBSHKWOFOGF-UHFFFAOYSA-L |
| IUPAC Name | disodium hydrate carbonate |
| SMILES | O.[Na+].[Na+].[O-]C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606386
|
Spectrum Chemical Mfg Cor
SO150500GM |
500 g | Poly Bottle |
Each for $548.96
|
|
||||
|
18606387
|
Spectrum Chemical Mfg Cor
SO15025KG |
2.5 kg | Poly Pail |
Each for $1,612.04
|
|
||||
|
18606388
|
Spectrum Chemical Mfg Cor
SO15012KG |
12 kg | Poly Pail |
Each for $2,995.21
|
|
||||
|
18606389
|
Spectrum Chemical Mfg Cor
SO15050KGBL |
50 kg | Fiber Drum |
Each for $6,470.18
|
|
||||