missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, Hydrous, NF, 99.5-100.5%, Spectrum™ Chemical

SO150, 5968-11-6, Na2CO3·H2O

Supplier:  Spectrum Chemical Mfg Cor SO15012KG

 View more versions of this product

Catalog No. 18606388


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Sodium Carbonate, Hydrous, NF is a pH regulator excipient. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

5968-11-6
12 kg
MQRJBSHKWOFOGF-UHFFFAOYSA-L
disodium hydrate carbonate
99.5 to 100.5%
Poly Pail
1
CH2Na2O4
O.[Na+].[Na+].[O-]C([O-])=O
124.00
NF

Chemical Identifiers

5968-11-6
124.00
disodium hydrate carbonate
CH2Na2O4
MQRJBSHKWOFOGF-UHFFFAOYSA-L
O.[Na+].[Na+].[O-]C([O-])=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions