missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, Hydrous, NF, 99.5-100.5%, Spectrum™ Chemical

Catalog No. 18606386
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606386 500 g Poly Bottle
18606387 2.5 kg Poly Pail
18606388 12 kg Poly Pail
18606389 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606386 Supplier Spectrum Chemical Mfg Cor Supplier No. SO150500GM
Only null left

SO150, 5968-11-6, Na2CO3·H2O

Spectrum™ Chemical Sodium Carbonate, Hydrous, NF is a pH regulator excipient. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5968-11-6
Molecular Formula CH2Na2O4
Molecular Weight (g/mol) 124.00
InChI Key MQRJBSHKWOFOGF-UHFFFAOYSA-L
IUPAC Name disodium hydrate carbonate
SMILES O.[Na+].[Na+].[O-]C([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99.5 to 100.5%
Grade NF
Packaging Poly Bottle