missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, 0.100 N (N/10), Ricca Chemical

Catalog No. p-2917021
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
718916 500 mL Natural Poly Bottle
718932 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 718916 Supplier Ricca Chemical Company Supplier No. 718916
Only null left

Chemical Identifiers

CAS 497-19-8 , 7732-18-5
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium carbonate
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Color Colorless
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.100N (N/10)
Packaging Natural Poly Bottle
pH 11 to 11.5
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Sodium Carbonate
ShelfLife 12 months