missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sodium cacodylate, 0.1M buffer soln., pH 7.0

Catalog No. p-7237393
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60344AD 50 mL
AAJ60344AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAJ60344AD Supplier Thermo Scientific™ Supplier No. J60344AD
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 6131-99-3 | C2H6AsNaO2 | 159.98 g/mol

Sodium cacodylate,is used as an arsinate compound for proteomics research. It is used as a buffer in electron microscopy.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 6131-99-3
Molecular Formula C2H6AsNaO2
Molecular Weight (g/mol) 159.98
MDL Number MFCD00149079
InChI Key IHQKEDIOMGYHEB-UHFFFAOYSA-M
IUPAC Name sodium dimethylarsinate
SMILES [Na+].C[As](C)([O-])=O

Specifications

Physical Form Liquid
Quantity 50 mL
ShelfLife 5 Years