missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bromate, Honeywell Fluka™
≥99%
$4199.88 - $4199.88
Chemical Identifiers
| CAS | 7789-38-0 |
|---|---|
| Molecular Formula | BrNaO3 |
| Molecular Weight (g/mol) | 150.891 |
| MDL Number | MFCD00003476 |
| InChI Key | XUXNAKZDHHEHPC-UHFFFAOYSA-M |
| Synonym | sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french |
| PubChem CID | 23668195 |
| ChEBI | CHEBI:75229 |
| IUPAC Name | sodium;bromate |
| SMILES | [O-]Br(=O)=O.[Na+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7789-38-0 | |
| 150.891 | |
| XUXNAKZDHHEHPC-UHFFFAOYSA-M | |
| 23668195 | |
| sodium;bromate |
| BrNaO3 | |
| MFCD00003476 | |
| sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french | |
| CHEBI:75229 | |
| [O-]Br(=O)=O.[Na+] |
Specifications
| 7789-38-0 | |
| BrNaO3 | |
| MFCD00003476 | |
| sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french | |
| [O-]Br(=O)=O.[Na+] | |
| 150.891 | |
| CHEBI:75229 | |
| ≥99% | |
| Sodium Bromate |
| 25 kg | |
| NaBrO3 | |
| 1494 | |
| XUXNAKZDHHEHPC-UHFFFAOYSA-M | |
| sodium;bromate | |
| 23668195 | |
| 150.89g/mol | |
| 3.339g/mL at 25°C |
Safety and Handling
P220-P261-P305 + P351 + P338
H272-H302-H315-H319-H335
EINECSNumber : 232-160-4
RTECSNumber : EF8750000