missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Borohydride, Crystalline Powder, Reagent, ACS, 98%, Spectrum™ Chemical
S1187, 16940-66-2, NaBH4
Supplier: Spectrum Chemical Mfg Cor S118750KGBL
Description
Spectrum™ Chemical Sodium Borohydride, Crystalline Powder, Reagent, ACS is an inorganic compound that is white in color and usually found as a powder. It is a reducing agent with versatility and is used as a synthetic reagent in the laboratory. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Specifications
| 16940-66-2 | |
| 50 kg | |
| MFCD00003518 | |
| [BH4-].[Na+] | |
| 37.83 | |
| ACS |
| 1 | |
| BH4Na | |
| YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
| sodium boranuide | |
| 98% | |
| Fiber Drum |
Chemical Identifiers
| 16940-66-2 | |
| 37.83 | |
| YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
| [BH4-].[Na+] |
| BH4Na | |
| MFCD00003518 | |
| sodium boranuide |