missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borohydride, Crystalline Powder, Reagent, ACS, 98%, Spectrum™ Chemical

Catalog No. p-7158191
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
40 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606365 25 g Amber Glass Bottle
18606366 100 g Amber Glass Bottle
18606367 500 g Amber Glass Bottle
18606368 2.5 kg Poly Pail
18606370 40 kg Fiber Drum
18606369 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18606365 Supplier Spectrum Chemical Mfg Cor Supplier No. S118725GM
Only null left

S1187, 16940-66-2, NaBH4

Spectrum™ Chemical Sodium Borohydride, Crystalline Powder, Reagent, ACS is an inorganic compound that is white in color and usually found as a powder. It is a reducing agent with versatility and is used as a synthetic reagent in the laboratory. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 98%
Grade ACS
Packaging Amber Glass Bottle