missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borate, Decahydrate, Powder, ACS, 99.5-105.0%, Spectrum™ Chemical

$368.32 - $5388.79

Chemical Identifiers

CAS 1303-96-4
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18613320
View Documents
Spectrum Chemical Mfg Cor
S1185500GM
500 g Poly Bottle
Each for $368.32
Only null left
 
18613321
View Documents
Spectrum Chemical Mfg Cor
S118525KG
2.5 kg Poly Bottle
Each for $873.94
Only null left
 
18613322
View Documents
Spectrum Chemical Mfg Cor
S118512KG
12 kg Poly Pail
Each for $2,290.15
Only null left
 
18613323
View Documents
Spectrum Chemical Mfg Cor
S118550KGBL
50 kg Fiber Drum
Each for $5,388.79
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1303-96-4

Specifications

1303-96-4
9.15 to 9.20
99.5 to 105.0%
Poly Bottle
1
500 g
ACS
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.