missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borate, Decahydrate, Powder, ACS, 99.5-105.0%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S118525KG

 View more versions of this product

Catalog No. 18613321


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1303-96-4
9.15 to 9.20
99.5 to 105.0%
Poly Bottle
1
2.5 kg
ACS

Chemical Identifiers

1303-96-4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.