missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bis(2-methoxyethoxy)aluminum hydride, 70 wt% sol. in toluene (ca. 3.5M)
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
| Color | Amber to Colorless |
| Boiling Point | 110.0°C |
| Linear Formula | [(CH3OCH2CH2O)2AlH2]Na |
| Fieser | 02,382; 03,260; 04,441; 05,596; 06,528; 07,327; 08,448; 12,440; 15,290 |
| Density | 1.0360g/mL |
| Solubility Information | Solubility in water: reacts. Other solubilities: soluble in aromatic hydrocarbons and ethers |
| Formula Weight | 202.16 |
| Percent Purity | 68.5 to 71.5% |
| Packaging | Glass bottle |
| Specific Gravity | 1.036 |
| Show More |
Safety and Handling
| Hazard Category | Danger |
RUO – Research Use Only