missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bicarbonate, puriss. p.a. ACS Reagent, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. Ph. Eur., powder,≥99.7%
Supplier: Honeywell Chemicals 3143750KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Sodium Bicarbonate | |
| 144-55-8 | |
| 50 kg | |
| NaHCO3 | |
| NONH for all modes of transport | |
| sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
| UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
| sodium;hydrogen carbonate | |
| 516892 | |
| 84.01g/mol | |
| ACS/puriss. p.a. |
| Reag. Ph. Eur. | |
| Powder | |
| CHNaO3 | |
| MFCD00003528 | |
| 4153970 | |
| Soluble in water, 50g/L | |
| [Na+].OC([O-])=O | |
| 84.01 | |
| CHEBI:32139 | |
| ≥99.7% |
Chemical Identifiers
| 144-55-8 | |
| 84.01 | |
| UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
| 516892 | |
| sodium;hydrogen carbonate |
| CHNaO3 | |
| MFCD00003528 | |
| sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
| CHEBI:32139 | |
| [Na+].OC([O-])=O |
Safety and Handling
EINECSNumber : 205-633-8
RTECSNumber : VZ0950000