missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate, puriss. p.a. ACS Reagent, Honeywell Fluka™

Catalog No. p-7072028
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
6 x 500 g
4 x 5 kg
50 kg
6 x 1 kg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002360 500 g
6002359 1 kg
60045953 6 x 500 g
6002362 5 kg
60045952 6 x 1 kg
60045951 4 x 5 kg
6002361 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 6002360 Supplier Honeywell Chemicals Supplier No. 31437500G
Only null left

Puriss. p.a., ACS Reagent, Reag. Ph. Eur., powder,≥99.7%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Physical Form Powder
Quantity 500 g
Linear Formula NaHCO3
UN Number NONH for all modes of transport
Beilstein 4153970
Solubility Information Soluble in water, 50g/L
Formula Weight 84.01g/mol
Percent Purity ≥99.7%
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium Bicarbonate, Reag. Ph. Eur.
EINECSNumber 205-633-8
RTECSNumber VZ0950000