missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Sodium Bicarbonate, Powder
Part of buffering system commonly used to maintain physiological pH 7.2 to 7.4 in culture environment
$414.26 - $414.26
Specifications
| Content And Storage | 15° to 30°C |
|---|---|
| Form | Powder |
| Product Type | Sodium Bicarbonate |
| Sterility | Sterile-filtered |
| Format | Plastic Bottle |
Description
Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.
- Molecular weight: 84.01
Specifications
| 15° to 30°C | |
| Sodium Bicarbonate | |
| Plastic Bottle |
| Powder | |
| Sterile-filtered | |
| 6.8 to 7.4 |