missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium bicarbonate, 1M buffer soln., pH 9.0

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

Supplier:  Thermo Scientific Chemicals J60066AP

 View more versions of this product

Catalog No. AAJ60066AP


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium bicarbonate buffer soln.
1 M
Liquid
CHNaO3
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium;hydrogen carbonate
516892
144-55-8
9.0
500 mL
MFCD00003528
[Na+].OC([O-])=O
84.01
CHEBI:32139

Chemical Identifiers

144-55-8
84.01
UIIMBOGNXHQVGW-UHFFFAOYSA-M
CHEBI:32139
[Na+].OC([O-])=O
CHNaO3
MFCD00003528
516892
sodium;hydrogen carbonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only