missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium bicarbonate, 1M buffer soln., pH 9.0

Catalog No. AAJ60066AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60066AK 250 mL
AAJ60066AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60066AK Supplier Thermo Scientific Chemicals Supplier No. J60066AK

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Concentration 1 M
pH 9.0
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material Sodium bicarbonate buffer soln.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only